Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbpl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144500.3 | NP_653083.2 | 12 - 13 | 1461 | 66 | |
GenBank mRNA | AK048537.1 | - | 12 - 13 | 1381 | 66 | |
AK145292.1 | - | 12 - 13 | 1493 | 66 | ||
AK166059.1 | - | 12 - 13 | 1440 | 66 | ||
AK170306.1 | - | 12 - 13 | 1445 | 66 | ||
AK170669.1 | - | 12 - 13 | 1462 | 66 | ||
AK220257.1 | - | 6 - 7 | 780 | 66 | ||
BC026804.1 | - | 12 - 13 | 1468 | 4159217 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |