Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lekr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037923.4 | NP_001033012.2 | 1 - 2 | 82 | 82 | |
NM_001166659.1 | NP_001160131.1 | 1 - 2 | 82 | 82 | ||
GenBank mRNA | AK137452.1 | - | 1 - 2 | 42 | 82 | |
CJ055787.1 | - | 1 - 2 | 77 | 82 | ||
CJ137734.1 | - | 1 - 2 | 83 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |