Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276292.1 | NP_001263221.1 | 15 - 16 | 1853 | 86 | |
NM_177327.5 | NP_796301.2 | 16 - 17 | 2015 | 86 | ||
NR_074089.1 | 15 - 16 | 1690 | 86 | |||
NR_074090.1 | 16 - 17 | 1912 | 86 | |||
GenBank mRNA | AK032232.1 | - | 15 - 16 | 1681 | 86 | |
AK033138.1 | - | 8 - 9 | 829 | 86 | ||
AK137588.1 | - | 16 - 17 | 1940 | 86 | ||
AK168567.1 | - | 16 - 17 | 1879 | 86 | ||
BC026829.1 | - | 15 - 16 | 1791 | 4209397 | 86 | |
BC150812.1 | - | 15 - 16 | 1850 | 9087723 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |