Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031168.1 | NP_112445.1 | 1 - 2 | 52 | 67 | |
GenBank mRNA | AK089780.1 | - | 1 - 2 | 71 | 67 | |
AK150440.1 | - | 1 - 2 | 71 | 67 | ||
BC132458.1 | - | 1 - 2 | 45 | 40130735 | 67 | |
BC138766.1 | - | 1 - 2 | 45 | 8861788 | 67 | |
BC145409.1 | - | 1 - 2 | 45 | 67 | ||
J03783.1 | - | 1 - 2 | 69 | 67 | ||
X06203.1 | - | 1 - 2 | 54 | 67 | ||
X54542.1 | - | 1 - 2 | 52 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |