Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133907.3 | NP_598668.1 | 10 - 11 | 1604 | 61 | |
GenBank mRNA | AK029973.1 | - | 10 - 11 | 1414 | 61 | |
AK045211.1 | - | 10 - 11 | 1402 | 61 | ||
AK046056.1 | - | 10 - 11 | 1605 | 61 | ||
AK122187.1 | - | 10 - 11 | 1539 | 61 | ||
BC059245.1 | - | 10 - 11 | 1419 | 61 | ||
BC120731.1 | - | 10 - 11 | 1351 | 40129654 | 61 | |
BC137626.1 | - | 10 - 11 | 1351 | 8860645 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |