Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yes1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205132.1 | NP_001192061.1 | 7 - 8 | 1084 | 88 | |
NM_001205133.1 | NP_001192062.1 | 7 - 8 | 1084 | 88 | ||
NM_009535.3 | NP_033561.1 | 7 - 8 | 1546 | 88 | ||
GenBank mRNA | AK035612.1 | - | 7 - 8 | 1533 | 88 | |
AK052478.1 | - | 7 - 8 | 1036 | 88 | ||
AK167420.1 | - | 7 - 8 | 1221 | 88 | ||
BC010594.1 | - | 7 - 8 | 920 | 3989783 | 88 | |
X67677.1 | - | 7 - 8 | 1473 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |