Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009316.1 | NP_033342.1 | 10 - 11 | 1234 | 79 | |
NM_172688.3 | NP_766276.1 | 10 - 11 | 1234 | 79 | ||
GenBank mRNA | AK046565.1 | - | 10 - 11 | 1200 | 79 | |
AK053911.1 | - | 10 - 11 | 1220 | 79 | ||
AK133642.1 | - | 4 - 5 | 361 | 79 | ||
AK137046.1 | - | 10 - 11 | 1235 | 79 | ||
AK159277.1 | - | 10 - 11 | 1198 | 79 | ||
BC006665.1 | - | 10 - 11 | 1225 | 3499247 | 79 | |
CV559615.1 | - | 3 - 4 | 290 | 79 | ||
D76446.1 | - | 10 - 11 | 1234 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |