Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272057.1 | NP_001258986.1 | 8 - 9 | 1035 | 116 | |
NM_001272058.1 | NP_001258987.1 | 8 - 9 | 1035 | 116 | ||
NM_001272059.1 | NP_001258988.1 | 8 - 9 | 1035 | 116 | ||
NM_007401.3 | NP_031427.2 | 8 - 9 | 1035 | 116 | ||
GenBank mRNA | AK016574.1 | - | 8 - 9 | 691 | 116 | |
AK029519.1 | - | 8 - 9 | 725 | 116 | ||
AK161262.1 | - | 8 - 9 | 1035 | 116 | ||
AK161415.1 | - | 5 - 6 | 413 | 116 | ||
AK161417.1 | - | 8 - 9 | 705 | 116 | ||
BC094237.1 | - | 8 - 9 | 732 | 30604517 | 116 | |
U22059.1 | - | 8 - 9 | 699 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |