Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032413.1 | NP_001027585.1 | 19 - 20 | 2632 | 129 | |
NM_001032414.1 | NP_001027586.1 | 18 - 19 | 2562 | 129 | ||
NM_001289600.1 | NP_001276529.1 | 18 - 19 | 2534 | 129 | ||
NM_001289601.1 | NP_001276530.1 | 20 - 21 | 2775 | 129 | ||
NM_028460.2 | NP_082736.1 | 20 - 21 | 2766 | 129 | ||
NM_152799.2 | NP_690012.2 | 20 - 21 | 2731 | 129 | ||
GenBank mRNA | AF440279.1 | - | 20 - 21 | 2756 | 129 | |
AF444274.1 | - | 18 - 19 | 2556 | 129 | ||
AK053551.1 | - | 19 - 20 | 2623 | 129 | ||
AK131151.1 | - | 4 - 5 | 849 | 129 | ||
AK141082.1 | - | 2529 | 129 | |||
AK161049.1 | - | 20 - 21 | 2773 | 129 | ||
BC042490.1 | - | 20 - 21 | 2491 | 4953584 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |