Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Veph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145820.3 | NP_665819.2 | 5 - 6 | 1077 | 133 | |
GenBank mRNA | AB085187.1 | - | 5 - 6 | 1059 | 133 | |
BC075649.1 | - | 5 - 6 | 1077 | 133 | ||
BC128271.1 | - | 4 - 5 | 784 | 40059997 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |