Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500003464.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003464.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003464.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003458.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003458.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003458.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003457.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003457.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003457.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003459.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003459.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003459.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003462.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003462.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003462.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003463.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003463.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003463.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003460.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003460.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003460.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003465.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003465.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003465.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003446.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003446.mm.1 |
NM_001081102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003446.mm.1 |
NM_001081102.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003446.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003446.mm.1 |
NM_001177884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003446.mm.1 |
NM_175231.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003446.mm.1 |
NM_175231.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003461.mm.1 |
TC0500000363.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500003461.mm.1 |
NM_001177884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500003461.mm.1 |
NM_001177884.1 |
Refseq |