Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012363.2 | NP_001012363.2 | 9 - 10 | 1128 | 89 | |
NM_001102414.1 | NP_001095884.1 | 9 - 10 | 1327 | 89 | ||
NM_001102415.1 | NP_001095885.1 | 7 - 8 | 1006 | 89 | ||
NM_145559.2 | NP_663534.1 | 8 - 9 | 1001 | 89 | ||
GenBank mRNA | AF469480.1 | - | 9 - 10 | 1331 | 89 | |
AF490463.1 | - | 7 - 8 | 831 | 89 | ||
AK033725.1 | - | 2 - 3 | 198 | 89 | ||
AK037093.1 | - | 8 - 9 | 992 | 89 | ||
AK156740.1 | - | 9 - 10 | 1231 | 89 | ||
AK172234.1 | - | 9 - 10 | 1231 | 89 | ||
AY776155.1 | - | 9 - 10 | 1107 | 89 | ||
BC006076.1 | - | 8 - 9 | 1001 | 3593581 | 89 | |
BC138213.1 | - | 10 - 11 | 1263 | 8861232 | 89 | |
BC145198.1 | - | 11 - 12 | 9053749 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |