Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400019258.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019258.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019258.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019262.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019262.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019262.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019264.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019264.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019264.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019268.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019268.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019268.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019259.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019259.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019259.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019270.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019270.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019270.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019260.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019260.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019260.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019271.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019271.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019271.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019267.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019267.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019267.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019261.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019261.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019261.mm.1 |
TC0400002372.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400019269.mm.1 |
NM_001034867.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019269.mm.1 |
NM_001034867.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400019269.mm.1 |
TC0400002372.mm.1 |
MTA TC |