Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024241.2 | NP_077203.2 | 3 - 4 | 953 | 141 | |
GenBank mRNA | AK016916.1 | - | 5 - 6 | 1151 | 141 | |
AK050683.1 | - | 3 - 4 | 939 | 141 | ||
AK135307.1 | - | 3 - 4 | 954 | 141 | ||
BC067395.1 | - | 3 - 4 | 945 | 30460089 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |