Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024241.2 | NP_077203.2 | 4 - 5 | 1055 | 106 | |
GenBank mRNA | AK016916.1 | - | 6 - 7 | 1253 | 106 | |
AK050683.1 | - | 4 - 5 | 1041 | 106 | ||
AK135307.1 | - | 4 - 5 | 1056 | 106 | ||
BC067395.1 | - | 4 - 5 | 1047 | 30460089 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |