Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175138.4 | NP_780347.2 | 9 - 10 | 1018 | 66 | |
GenBank mRNA | AK030213.1 | - | 9 - 10 | 952 | 66 | |
AK047808.1 | - | 9 - 10 | 1019 | 66 | ||
BC138785.1 | - | 9 - 10 | 963 | 8861807 | 66 | |
BC138787.1 | - | 9 - 10 | 963 | 8861809 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |