Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013770.3 | NP_001013792.2 | 3 - 4 | 668 | 86 | |
GenBank mRNA | AK139790.1 | - | 3 - 4 | 669 | 86 | |
AK170332.1 | - | 3 - 4 | 602 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |