Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025700.2 | NP_079976.1 | 11 - 12 | 1473 | 155 | |
GenBank mRNA | AK004286.1 | - | 2 - 3 | 154 | 155 | |
AK014332.1 | - | 11 - 12 | 1398 | 155 | ||
BC030869.1 | - | 11 - 12 | 1435 | 155 | ||
BC052762.1 | - | 11 - 12 | 1473 | 6510389 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |