Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177815.3 | NP_808483.2 | 5 - 6 | 591 | 73 | |
GenBank mRNA | BC082539.1 | - | 5 - 6 | 559 | 73 | |
BC094336.1 | - | 5 - 6 | 559 | 73 | ||
BC141275.1 | - | 5 - 6 | 585 | 9055887 | 73 | |
BC145606.1 | - | 5 - 6 | 585 | 9054172 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |