Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177815.3 | NP_808483.2 | 8 - 9 | 859 | 88 | |
GenBank mRNA | AK184779.1 | - | 3 - 4 | 260 | 89 | |
BC094336.1 | - | 8 - 9 | 827 | 88 | ||
BC141275.1 | - | 8 - 9 | 853 | 9055887 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |