Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008872.2 | NP_032898.2 | 8 - 9 | 903 | 54 | |
GenBank mRNA | AK135857.1 | - | 8 - 9 | 894 | 54 | |
AK135965.1 | - | 8 - 9 | 895 | 54 | ||
AK141665.1 | - | 8 - 9 | 892 | 54 | ||
AK159620.1 | - | 8 - 9 | 895 | 54 | ||
AK166639.1 | - | 8 - 9 | 893 | 54 | ||
BC011256.1 | - | 8 - 9 | 900 | 4038404 | 54 | |
BC057967.1 | - | 8 - 9 | 883 | 4979664 | 54 | |
BC061508.1 | - | 8 - 9 | 884 | 5352018 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |