Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145952.3 | NP_666064.3 | 11 - 12 | 1898 | 154 | |
GenBank mRNA | AK172979.1 | - | 11 - 12 | 1892 | 154 | |
BC033574.1 | - | 2 - 3 | 200 | 154 | ||
BC138056.1 | - | 11 - 12 | 1794 | 8861075 | 154 | |
BC138057.1 | - | 11 - 12 | 1794 | 8861076 | 154 | |
BC145129.1 | - | 9 - 10 | 1569 | 9053677 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |