Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025652.3 | NP_079928.2 | 3 - 4 | 516 | 70 | |
GenBank mRNA | AK008247.2 | - | 3 - 4 | 483 | 70 | |
AK011845.1 | - | 3 - 4 | 465 | 70 | ||
AK012797.1 | - | 3 - 4 | 463 | 70 | ||
AK017234.1 | - | 3 - 4 | 410 | 70 | ||
AK143627.1 | - | 3 - 4 | 468 | 70 | ||
BC032292.1 | - | 2 - 3 | 171 | 5374268 | 70 | |
BC057966.1 | - | 2 - 3 | 728 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |