Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013691.2 | NP_038719.2 | 11 - 12 | 1387 | 83 | |
GenBank mRNA | BC053023.1 | - | 11 - 12 | 1341 | 6401682 | 83 |
BC060104.1 | - | 10 - 11 | 2137 | 83 | ||
L04302.1 | - | 11 - 12 | 1367 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |