Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109661.1 | NP_001103131.1 | 1 - 2 | 150 | 83 | |
GenBank mRNA | BC131642.1 | - | 1 - 2 | 58 | 40109815 | 83 |
CJ092609.1 | - | 2 - 3 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |