Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109661.1 | NP_001103131.1 | 2 - 3 | 411 | 73 | |
GenBank mRNA | BC131642.1 | - | 2 - 3 | 319 | 40109815 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |