Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007641.5 | NP_031667.1 | 2 - 3 | 258 | 65 | |
GenBank mRNA | AK017903.1 | - | 2 - 3 | 259 | 65 | |
AK041428.1 | - | 2 - 3 | 257 | 65 | ||
AK080040.1 | - | 2 - 3 | 224 | 65 | ||
AK080148.1 | - | 256 | 65 | |||
AK137469.1 | - | 2 - 3 | 247 | 65 | ||
AK143824.1 | - | 2 - 3 | 253 | 65 | ||
AK156033.1 | - | 257 | 65 | |||
AK166571.1 | - | 2 - 3 | 351 | 65 | ||
AK166659.1 | - | 2 - 3 | 351 | 65 | ||
AK171631.1 | - | 2 - 3 | 287 | 65 | ||
AK171638.1 | - | 2 - 3 | 210 | 65 | ||
AK171659.1 | - | 2 - 3 | 243 | 65 | ||
BC028322.1 | - | 2 - 3 | 239 | 4503841 | 65 | |
CT010253.1 | - | 1 - 2 | 139 | 65 | ||
M62541.1 | - | 316 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |