Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013517.4 | NP_038545.1 | 2 - 3 | 196 | 67 | |
GenBank mRNA | AK080159.1 | - | 2 - 3 | 197 | 67 | |
AK156654.1 | - | 2 - 3 | 199 | 67 | ||
AK172591.1 | - | 2 - 3 | 199 | 67 | ||
BC119164.1 | - | 1 - 2 | 63 | 8733913 | 67 | |
BC119190.1 | - | 1 - 2 | 63 | 8733939 | 67 | |
M34163.1 | - | 2 - 3 | 198 | 67 | ||
M99371.1 | - | 2 - 3 | 188 | 67 | ||
S67072.1 | - | 1 - 2 | 47 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |