Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016979.1 | NP_058675.1 | 6 - 7 | 1126 | 51 | |
GenBank mRNA | AJ238004.1 | - | 6 - 7 | 1009 | 51 | |
AK034681.1 | - | 6 - 7 | 1445 | 51 | ||
AK037141.1 | - | 6 - 7 | 1120 | 51 | ||
AK081548.1 | - | 6 - 7 | 1106 | 51 | ||
AK139510.1 | - | 6 - 7 | 1134 | 51 | ||
AK149937.1 | - | 6 - 7 | 1104 | 51 | ||
AK150588.1 | - | 6 - 7 | 1088 | 51 | ||
AK152166.1 | - | 6 - 7 | 922 | 51 | ||
AK154447.1 | - | 6 - 7 | 1106 | 51 | ||
AK169322.1 | - | 6 - 7 | 1077 | 51 | ||
BC006875.1 | - | 6 - 7 | 1126 | 3596013 | 51 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |