Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017471.2 | NP_059499.1 | 4 - 5 | 408 | 141 | |
GenBank mRNA | AF005204.1 | - | 4 - 5 | 412 | 141 | |
AK137501.1 | - | 4 - 5 | 435 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |