Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159573.1 | NP_001153045.1 | 4 - 5 | 427 | 81 | |
NM_001159574.1 | NP_001153046.1 | 4 - 5 | 427 | 81 | ||
NM_024183.5 | NP_077145.2 | 4 - 5 | 427 | 81 | ||
GenBank mRNA | AK005061.1 | - | 4 - 5 | 396 | 81 | |
AK008561.1 | - | 4 - 5 | 399 | 81 | ||
AK049672.1 | - | 4 - 5 | 413 | 81 | ||
AK164105.1 | - | 4 - 5 | 393 | 81 | ||
AK168464.1 | - | 4 - 5 | 428 | 81 | ||
BC003263.1 | - | 4 - 5 | 385 | 3499130 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |