Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007154.3 | NP_001007155.1 | 9 - 10 | 1367 | 67 | |
NM_001177789.1 | NP_001171260.1 | 9 - 10 | 1585 | 67 | ||
NM_001177790.1 | NP_001171261.1 | 9 - 10 | 1582 | 67 | ||
NM_001177791.1 | NP_001171262.1 | 9 - 10 | 1419 | 67 | ||
NM_028806.2 | NP_083082.1 | 9 - 10 | 1824 | 67 | ||
GenBank mRNA | AJ632220.1 | - | 9 - 10 | 1382 | 67 | |
AJ632221.1 | - | 9 - 10 | 1420 | 67 | ||
AJ632222.1 | - | 9 - 10 | 1367 | 67 | ||
AJ632223.1 | - | 8 - 9 | 1257 | 67 | ||
AK005136.1 | - | 9 - 10 | 1389 | 67 | ||
AK032242.1 | - | 9 - 10 | 1591 | 67 | ||
AK038449.1 | - | 9 - 10 | 1335 | 67 | ||
AK039213.1 | - | 9 - 10 | 1374 | 67 | ||
AK135144.1 | - | 9 - 10 | 1825 | 67 | ||
AK220554.1 | - | 9 - 10 | 1582 | 67 | ||
AY501387.1 | - | 9 - 10 | 1531 | 67 | ||
BC058621.1 | - | 9 - 10 | 1740 | 5708186 | 67 | |
BC059869.1 | - | 9 - 10 | 1603 | 6809310 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |