Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037722.2 | NP_001032811.2 | 10 - 11 | 1131 | 88 | |
NM_009614.2 | NP_033744.1 | 10 - 11 | 1131 | 88 | ||
GenBank mRNA | AF006196.1 | - | 10 - 11 | 1092 | 88 | |
AK039384.1 | - | 10 - 11 | 1111 | 88 | ||
AK048901.1 | - | 10 - 11 | 1118 | 88 | ||
AK142520.1 | - | 10 - 11 | 1113 | 88 | ||
AK149684.1 | - | 10 - 11 | 1112 | 88 | ||
AK149796.1 | - | 1104 | 88 | |||
AK151804.1 | - | 10 - 11 | 1067 | 88 | ||
AK152725.1 | - | 10 - 11 | 1067 | 88 | ||
AK152954.1 | - | 10 - 11 | 1068 | 88 | ||
AK152973.1 | - | 10 - 11 | 1068 | 88 | ||
AK170101.1 | - | 10 - 11 | 1097 | 88 | ||
AK170365.1 | - | 10 - 11 | 1111 | 88 | ||
BC009132.1 | - | 10 - 11 | 1080 | 3152945 | 88 | |
BC057909.1 | - | 9 - 10 | 1273 | 88 | ||
EF506571.1 | - | 4 - 5 | 392 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |