Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039466.1 | NP_001034555.1 | 2 - 3 | 341 | 71 | |
NM_183037.2 | NP_898858.1 | 2 - 3 | 444 | 71 | ||
NR_121683.1 | 2 - 3 | 574 | 71 | |||
GenBank mRNA | AK132368.1 | - | 2 - 3 | 310 | 71 | |
AK170904.1 | - | 2 - 3 | 342 | 71 | ||
AY251388.1 | - | 2 - 3 | 324 | 71 | ||
BC065049.1 | - | 2 - 3 | 487 | 6418851 | 71 | |
BC082299.1 | - | 2 - 3 | 444 | 71 | ||
CA315205.1 | - | 2 - 3 | 574 | 71 | ||
CA320885.1 | - | 2 - 3 | 444 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |