Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10605031 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600120 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600119 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600121 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605033 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600117 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605032 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600149 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605030 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10605029 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10600118 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10600148 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10600148 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10605028 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10600116 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10600116 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10605028 |
- |
- |
Mouse Genome 430 2.0 Array |
1429745_at |
- |
- |
Mouse Genome 430 2.0 Array |
1429745_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0X00004240.mm.1 |
NM_001025383.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004240.mm.1 |
NM_001025383.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004240.mm.1 |
TC0X00000674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004243.mm.1 |
NM_001025383.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004243.mm.1 |
NM_001025383.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004243.mm.1 |
TC0X00000674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004247.mm.1 |
NM_001025383.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004247.mm.1 |
NM_001025383.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004247.mm.1 |
TC0X00000674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014955.mm.1 |
NM_001025384.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014955.mm.1 |
NM_001025384.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014955.mm.1 |
TC0X00002420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00004242.mm.1 |
NM_001025383.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004242.mm.1 |
NM_001025383.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00004242.mm.1 |
TC0X00000674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014953.mm.1 |
NM_001025384.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014953.mm.1 |
NM_001025384.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014953.mm.1 |
TC0X00002420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014954.mm.1 |
NM_001025384.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014954.mm.1 |
NM_001025384.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014954.mm.1 |
TC0X00002420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014956.mm.1 |
NM_001025384.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014956.mm.1 |
NM_001025384.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014956.mm.1 |
TC0X00002420.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00014951.mm.1 |
NM_001025384.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014951.mm.1 |
NM_001025384.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00014951.mm.1 |
TC0X00002420.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1429745_at |
- |
- |
Mouse Expression Set 430 |
1429745_PM_at |
- |
- |
Mouse Expression Set 430 |
1429745_PM_at |
- |
- |
Mouse Expression Set 430 |
1429745_at |
- |
- |
Murine Genome U74v2 Set |
169961_PM_at |
- |
- |
Murine Genome U74v2 Set |
169961_at |
- |
- |
Murine Genome U74v2 Set |
169961_PM_at |
- |
- |
Murine Genome U74v2 Set |
169961_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1429745_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1429745_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1429745_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1429745_PM_at |
- |
- |