Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122754.2 | NP_001116226.1 | 13 - 14 | 1435 | 160 | |
NM_001289494.1 | NP_001276423.1 | 12 - 13 | 1369 | 160 | ||
NM_001289495.1 | NP_001276424.1 | 13 - 14 | 1452 | 160 | ||
NM_013698.3 | NP_038726.2 | 12 - 13 | 1519 | 160 | ||
GenBank mRNA | AK088209.1 | - | 12 - 13 | 1366 | 160 | |
AK138786.1 | - | 13 - 14 | 1449 | 160 | ||
BC120552.1 | - | 12 - 13 | 1422 | 40129475 | 160 | |
BC137710.1 | - | 12 - 13 | 1424 | 8860729 | 160 | |
D43963.1 | - | 12 - 13 | 1519 | 160 | ||
D83201.1 | - | 12 - 13 | 1459 | 160 | ||
D83202.1 | - | 12 - 13 | 1622 | 160 | ||
U16145.1 | - | 13 - 14 | 1400 | 160 | ||
U19607.1 | - | 13 - 14 | 1435 | 160 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |