Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122754.2 | NP_001116226.1 | 5 - 6 | 533 | 72 | |
NM_001289495.1 | NP_001276424.1 | 5 - 6 | 550 | 72 | ||
NM_013698.3 | NP_038726.2 | 4 - 5 | 617 | 72 | ||
GenBank mRNA | AK138069.1 | - | 5 - 6 | 530 | 72 | |
AK138786.1 | - | 5 - 6 | 547 | 72 | ||
BC120552.1 | - | 4 - 5 | 520 | 40129475 | 72 | |
BC137710.1 | - | 4 - 5 | 522 | 8860729 | 72 | |
D43963.1 | - | 4 - 5 | 617 | 72 | ||
D83201.1 | - | 4 - 5 | 557 | 72 | ||
D83202.1 | - | 4 - 5 | 720 | 72 | ||
U16145.1 | - | 5 - 6 | 498 | 72 | ||
U19607.1 | - | 5 - 6 | 533 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |