Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253719.1 | NP_001240648.1 | 8 - 9 | 1185 | 64 | |
NM_178673.4 | NP_848788.2 | 8 - 9 | 1445 | 64 | ||
GenBank mRNA | AF374460.1 | - | 8 - 9 | 1419 | 64 | |
AK033652.1 | - | 8 - 9 | 1436 | 64 | ||
AK046070.1 | - | 8 - 9 | 1186 | 64 | ||
AK081152.1 | - | 8 - 9 | 1436 | 64 | ||
AK138388.1 | - | 8 - 9 | 1737 | 64 | ||
BC075640.1 | - | 8 - 9 | 1408 | 30545529 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |