Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144945.2 | NP_659194.1 | 2 - 3 | 434 | 110 | |
GenBank mRNA | AJ487515.1 | - | 2 - 3 | 457 | 110 | |
AK122570.1 | - | 2 - 3 | 399 | 110 | ||
AY841361.1 | - | 2 - 3 | 259 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |