Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027009.2 | NP_081285.1 | 5 - 6 | 623 | 71 | |
GenBank mRNA | AK042393.1 | - | 5 - 6 | 624 | 71 | |
AK145126.1 | - | 5 - 6 | 650 | 71 | ||
AK165891.1 | - | 5 - 6 | 618 | 71 | ||
AK167047.1 | - | 5 - 6 | 624 | 71 | ||
AK209135.1 | - | 2 - 3 | 200 | 71 | ||
BC026795.1 | - | 5 - 6 | 594 | 4015134 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |