Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166457.1 | NP_001159929.1 | 1 - 2 | 215 | 86 | |
NM_001166459.1 | NP_001159931.1 | 1 - 2 | 215 | 86 | ||
NM_145956.4 | NP_666068.1 | 1 - 2 | 215 | 86 | ||
GenBank mRNA | AK011876.1 | - | 1 - 2 | 132 | 86 | |
AK030736.1 | - | 1 - 2 | 145 | 86 | ||
AK038919.1 | - | 1 - 2 | 133 | 86 | ||
AK149844.1 | - | 1 - 2 | 147 | 86 | ||
BC021313.1 | - | 1 - 2 | 162 | 5026715 | 86 | |
BC048179.1 | - | 1 - 2 | 129 | 6825974 | 86 | |
S68022.1 | - | 1 - 2 | 157 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |