Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF099067.1 | - | 10 - 11 | 1864 | 72 | |
AJ223071.1 | - | 10 - 11 | 1954 | 72 | ||
AK078809.1 | - | 9 - 10 | 1832 | 72 | ||
AK132803.1 | - | 9 - 10 | 1748 | 72 | ||
AK149584.1 | - | 9 - 10 | 1748 | 72 | ||
BC057939.1 | - | 10 - 11 | 1866 | 5325558 | 72 | |
Y09234.1 | - | 9 - 10 | 1810 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |