Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011103.3 | NP_035233.1 | 2 - 3 | 350 | 69 | |
GenBank mRNA | AB011812.1 | - | 1 - 2 | 113 | 69 | |
AF251036.1 | - | 1 - 2 | 113 | 69 | ||
AK144231.1 | - | 2 - 3 | 355 | 69 | ||
AK171476.1 | - | 2 - 3 | 351 | 69 | ||
AY545076.1 | - | 1 - 2 | 113 | 69 | ||
BC051416.1 | - | 2 - 3 | 314 | 6515302 | 69 | |
HQ840432.1 | - | 1 - 2 | 113 | 69 | ||
M69042.1 | - | 2 - 3 | 345 | 69 | ||
X60304.1 | - | 1 - 2 | 126 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |