Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB073078.1 | - | 7 - 8 | 916 | 107 | |
AB073080.1 | - | 8 - 9 | 988 | 107 | ||
AK050922.1 | - | 8 - 9 | 1017 | 107 | ||
AK142379.1 | - | 7 - 8 | 899 | 107 | ||
BC145176.1 | - | 6 - 7 | 807 | 9053727 | 107 | |
BC145850.1 | - | 7 - 8 | 879 | 40131153 | 107 | |
S66169.1 | - | 4 - 5 | 301 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |