Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153592.2 | NP_705820.1 | 8 - 9 | 679 | 62 | |
GenBank mRNA | AK030557.1 | - | 8 - 9 | 623 | 62 | |
BC036333.1 | - | 8 - 9 | 644 | 3590904 | 62 | |
BC038374.1 | - | 8 - 9 | 619 | 5355446 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |