Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Got1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029674.1 | NP_083950.1 | 2 - 3 | 355 | 61 | |
GenBank mRNA | AK006984.1 | - | 2 - 3 | 342 | 61 | |
BC052754.1 | - | 2 - 3 | 355 | 6533656 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |