Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_001165991.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_001165991.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_001165992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_001165992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_001165995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_001165995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_029702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NM_029702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NR_028585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
NR_028585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046457.mm.1 |
TC0200005443.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NM_001165991.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NM_001165991.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NM_001165992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NM_001165992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NM_029702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NM_029702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NR_028585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
NR_028585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046444.mm.1 |
TC0200005443.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_001165991.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_001165991.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_001165992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_001165992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_001165995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_001165995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_029702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NM_029702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NR_028585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
NR_028585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046450.mm.1 |
TC0200005443.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_001165991.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_001165991.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_001165992.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_001165992.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_001165995.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_001165995.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_029702.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NM_029702.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NR_028585.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
NR_028585.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046449.mm.1 |
TC0200005443.mm.1 |
MTA TC |