Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183161.3 | NP_898984.3 | 1 - 2 | 108 | 52 | |
GenBank mRNA | BC019537.1 | - | 1 - 2 | 68 | 52 | |
BG087579.2 | - | 1 - 2 | 108 | 52 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |