Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_001083328.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_001083328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_001083329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_001083329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_028422.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_028422.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_181424.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
NM_181424.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022705.mm.1 |
TC0200002781.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_001083328.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_001083328.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_001083329.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_001083329.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_028422.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_028422.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_181424.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
NM_181424.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200022708.mm.1 |
TC0200002781.mm.1 |
MTA TC |