Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gid8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289651.1 | NP_001276580.1 | 1 - 2 | 155 | 104 | |
GenBank mRNA | BC052539.1 | - | 1 - 2 | 153 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |